N4-Acetylcytosine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-(2-Oxo-1,2-dihydropyrimidin-4-yl)acetamide
A287573-1000G
A287573-100G
A287573-10G
A287573-25G
A287573-500G
A287573-5G
77277-104EA
135
USD
77277-104
77277-106
77277-108
77277-110
77277-112
77277-114
N4-Acetylcytosine 98%
N4-Acetylcytosine
Caution: for Research Use Only
Formula:
C₆H₇N₃O₂ |
MDL Number:
MFCD00134466 CAS Number: 14631-20-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...