3-Chloro-4-((3-fluorobenzyl)oxy)aniline 98%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-4-(3-fluorobenzyloxy)aniline

A266141-25G A266141-5G A266141-100G A266141-10G A266141-1G
103812-786EA 5 USD
103812-786 103852-686 77092-386 77092-388 77276-690 77276-692 77304-948
3-Chloro-4-((3-fluorobenzyl)oxy)aniline 98%
3-Chloro-4-((3-fluorobenzyl)oxy)aniline

Caution: for Research Use Only

Formula: C₁₃H₁₁ClFNO
Boiling Pt: 387.9 °C (760 mm Hg)
Flash Pt: 188.4 °C
Storage Temperature: Ambient
MDL Number: MFCD06809822
CAS Number: 202197-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR