3-Methoxybenzenesulfonyl chloride 97%

Supplier: Ambeed
Danger

Synonyms: 3-Methoxybenzene-1-sulfonyl chloride, 3-Methoxybenzene-1-sulphonyl chloride

A258727-1G A258727-250MG A258727-25G A258727-5G
77276-492EA 22 USD
77276-492 77276-494 77276-496 77276-498
3-Methoxybenzenesulfonyl chloride 97%
3-Methoxybenzenesulphonyl chloride

Caution: for Research Use Only

Formula: C₇H₇ClO₃S
MW: 206.65 g/mol
MDL Number: MFCD01318155
CAS Number: 10130-74-2
UN: 3265
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR