1-Boc-4-(aminomethyl)piperidine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(Aminomethyl)-1-tert-butoxycarbonylpiperidine, tert-Butyl 4-(aminomethyl)piperidine-1-carboxylate, tert-Butyl 4-(Aminomethyl)-1-piperidinecarboxylate, tert-Butyl 4-(aminomethyl)tetrahydropyridine-1(2H)- carboxylate, 4-(Aminomethyl)-1-Boc-piperidine, 4-(Aminomethyl)-1-piperidinecarboxylic acid tert-Butyl ester, 1-Boc-4-(Aminomethyl)piperidine
A254811-25G
A254811-5G
A254811-100G
A254811-10G
A254811-500G
A254811-1G
103812-168EA
9
USD
103812-168
103852-020
77091-468
77091-470
77276-396
77276-398
77276-400
77304-640
1-Boc-4-(aminomethyl)piperidine 97%
1-Boc-4-(aminomethyl)piperidine
Caution: for Research Use Only
Formula:
C₁₁H₂₂N₂O₂ Boiling Pt: 237…238 °C (760 mm Hg) Melting Pt: 102…106 °C Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD01076207 CAS Number: 144222-22-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...