1-Boc-4-(aminomethyl)piperidine 97%

Supplier: Ambeed
Warning

Synonyms: 4-(Aminomethyl)-1-tert-butoxycarbonylpiperidine, tert-Butyl 4-(aminomethyl)piperidine-1-carboxylate, tert-Butyl 4-(Aminomethyl)-1-piperidinecarboxylate, tert-Butyl 4-(aminomethyl)tetrahydropyridine-1(2H)- carboxylate, 4-(Aminomethyl)-1-Boc-piperidine, 4-(Aminomethyl)-1-piperidinecarboxylic acid tert-Butyl ester, 1-Boc-4-(Aminomethyl)piperidine

A254811-25G A254811-5G A254811-100G A254811-10G A254811-500G A254811-1G
103812-168EA 9 USD
103812-168 103852-020 77091-468 77091-470 77276-396 77276-398 77276-400 77304-640
1-Boc-4-(aminomethyl)piperidine 97%
1-Boc-4-(aminomethyl)piperidine

Caution: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
Boiling Pt: 237…238 °C (760 mm Hg)
Melting Pt: 102…106 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD01076207
CAS Number: 144222-22-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR