3-Bromo-4-hydroxybenzoic acid 98%
Supplier: Ambeed
|
Warning
|
A239630-100G
A239630-1G
A239630-25G
A239630-5G
77275-978EA
129
USD
77275-978
77275-980
77275-982
77275-984
3-Bromo-4-hydroxybenzoic acid 98%
3-Bromo-4-hydroxybenzoic acid
Caution: for Research Use Only
Formula:
C₇H₅BrO₃ MW: 217.02 |
MDL Number:
MFCD00017547 CAS Number: 14348-41-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...