3-Bromo-4-hydroxybenzoic acid 98%

Supplier: Ambeed
Warning

A239630-100G A239630-1G A239630-25G A239630-5G
77275-978EA 129 USD
77275-978 77275-980 77275-982 77275-984
3-Bromo-4-hydroxybenzoic acid 98%
3-Bromo-4-hydroxybenzoic acid

Caution: for Research Use Only

Formula: C₇H₅BrO₃
MW: 217.02
MDL Number: MFCD00017547
CAS Number: 14348-41-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR