2',3',5'-Tri-O-acetylguanosine 95%

Supplier: Ambeed
Warning

A218267-1G A218267-25G A218267-5G A218267-100G
77275-418EA 17.01 USD
77275-418 77275-420 77275-422 77748-346
2',3',5'-Tri-O-acetylguanosine 95%
2',3',5'-Tri-O-acetylguanosine

Caution: for Research Use Only

Formula: C₁₆H₁₉N₅O₈
MDL Number: MFCD00057054
CAS Number: 6979-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR