4-Bromomethyl-2-cyanobiphenyl 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
4'-(Bromomethyl)-[1,1'-biphenyl]-2-carbonitrile
A215394-1000G
A215394-100G
A215394-10G
A215394-500G
A215394-5G
77275-292EA
165
USD
77275-292
77275-294
77275-296
77275-298
77275-300
4-Bromomethyl-2-cyanobiphenyl 95%
4-Bromomethyl-2-cyanobiphenyl
Caution: for Research Use Only
Formula:
C₁₄H₁₀BrN MW: 272.14 g/mol |
MDL Number:
MFCD00671503 CAS Number: 114772-54-2 UN: 3261 ADR: 8 Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...