Indole-3-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1H-Indole-3-carboxylic acid
A188858-25G
A188858-100G
A188858-100MG
A188858-10G
A188858-500G
103846-730EA
6
USD
103846-730
77084-482
77274-368
77274-370
77274-372
77274-374
Indole-3-carboxylic acid 98%
Indole-3-carboxylic acid
Caution: for Research Use Only
Formula:
C₉H₇NO₂ Boiling Pt: 419.6 °C (760 mm Hg) Melting Pt: 232…240 °C Flash Pt: 207.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005624 CAS Number: 771-50-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...