(R)-(+)-2-Pyrrolidinone-5-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
D-Pyr-OH, (R)-(+)-2-Pyrrolidone-5-carboxylic acid
A167373-25G
A167373-100G
A167373-10G
A167373-5G
103844-438EA
5
USD
103844-438
77081-450
77273-578
77273-580
77300-892
(R)-(+)-2-Pyrrolidinone-5-carboxylic acid 98%
(R)-(+)-2-Pyrrolidinone-5-carboxylic acid
Caution: for Research Use Only
Formula:
C₅H₇NO₃ Boiling Pt: 453.1 °C (760 mm Hg) Melting Pt: 155…162 °C Flash Pt: 227.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066212 CAS Number: 4042-36-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...