3-Aminobutan-1-ol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Amino-butan-1-ol
A161784-100G
A161784-10G
A161784-500G
A161784-1G
A161784-25G
A161784-5G
103805-538EA
6
USD
103805-538
103843-780
77273-332
77273-334
77273-336
77319-792
77319-794
77319-796
3-Aminobutan-1-ol 97%
3-Aminobutan-1-ol
Caution: for Research Use Only
Formula:
C₄H₁₁NO Boiling Pt: 168.3 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD03412694 CAS Number: 2867-59-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...