Nɑ-(tert-Butoxycarbonyl)-N-ε-carbobenzoxy-L-lysine 98%

Supplier: Ambeed
Warning

A156665-100G A156665-500G A156665-5G A156665-25G A156665-1000G
103804-910EA 6 USD
103804-910 103843-112 77273-148 77273-150 77273-152 77300-206 77318-908 77748-067
Nɑ-(tert-Butoxycarbonyl)-N-ε-carbobenzoxy-L-lysine 98%
Nalpha-(tert-Butoxycarbonyl)-N-epsilon-carbobenzoxy-L-lysine

Caution: for Research Use Only

Formula: C₁₉H₂₈N₂O₆
Boiling Pt: 587 °C (760 mm Hg)
Flash Pt: 308.8 °C
Storage Temperature: Freezer
MDL Number: MFCD00065584
CAS Number: 2389-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR