2'-Fluoro-2'-deoxycytidine 95%
Supplier: Ambeed
|
Warning
|
A148637-1G
A148637-250MG
A148637-5G
77272-882EA
27
USD
77272-882
77272-884
77272-886
2'-Fluoro-2'-deoxycytidine 95%
2'-Fluoro-2'-deoxycytidine
Caution: for Research Use Only
Formula:
C₉H₁₂FN₃O₄ |
MDL Number:
MFCD00057445 CAS Number: 10212-20-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...