3-Hydroxypropiononitrile 97%

Supplier: Ambeed
Warning

A141843-100G A141843-25G A141843-500G
77272-492EA 19 USD
77272-492 77272-494 77272-496
3-Hydroxypropiononitrile 97%
3-Hydroxypropiononitrile

Caution: for Research Use Only

Formula: C₃H₅NO
MDL Number: MFCD00002826
CAS Number: 109-78-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR