2'-Fluoro-4'-methoxyacetophenone 98%

Supplier: Ambeed
Warning

A121223-100G A121223-25G A121223-10G
103830-866EA 15 USD
103830-866 77271-506 77271-508 77747-880
2'-Fluoro-4'-methoxyacetophenone 98%
2'-Fluoro-4'-methoxyacetophenone

Caution: for Research Use Only

Formula: C₉H₉FO₂
Melting Pt: 52…54 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00042290
CAS Number: 74457-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR