Adenine hydrochloride 98%

Supplier: Ambeed
Warning

A114669-100G A114669-100MG A114669-25G A114669-500G
77271-136EA 51.01 USD
77271-136 77271-138 77271-140 77271-142
Adenine hydrochloride 98%
Adenine hydrochloride

Caution: for Research Use Only

Formula: C₅H₅N₅·HCl
MDL Number: MFCD00150864
CAS Number: 2922-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR