MicroSeal® NW Antistatic 100% Polyester Cleanroom Wipes
Supplier: Berkshire
MSNW090910
77270-042CS 544.5 USD
77270-042
MicroSeal® NW Antistatic 100% Polyester Cleanroom Wipes
Wipes Cleanroom Wipes Dry Cleanroom Wipes
MicroSeal® NW antistatic is a sealed edge knitted polyester wiper. A conductive fiber is knitted into the material to provide increased control over static.
  • Chemically compatible with common cleaning and disinfecting solutions
  • Double bagged
  • Ultrasonically sealed edge for extremely low fiber release
  • High abrasion resistance
  • Lot traceability
Laundered and packaged in Berkshire’s ISO Class 4 cleanroom.
Certifications: Cleanroom Class 1 - 100; EU Grade B/C/D.