Stainless Steel Multi-Prong Tool Holder for Stainless Steel LocBoard®
Supplier: Triton Products
66660
77269-318EA 14.43 USD
77269-318
Stainless Steel Multi-Prong Tool Holder for Stainless Steel LocBoard®
Wall Hooks
The stainless steel LocHook® is ideal for food, medical, healthcare, restaurant, marine and commercial applications as well as any clean room or harsh environment that must meet stringent requirements.
  • Ideal to hang box wrenches, screw drivers or any tools with a round hole that will fit over the prongs
  • Easily repositioned to accommodate changing storage needs with no damage to the stainless steel LocBoard
  • Won't rust or corrode in any environment, even ones with extreme temperatures
Having clean, easily accessible and versatile storage and organizational solutions is an issue that every business encounters. The food and healthcare industries must meet stringent requirements and The stainless steel pegboard storage from Triton products provides the ideal answer. Two innovative best-selling products, the LocBoard and the LocHook, are now available in stainless steel. They provide a commercial grade heavy-duty pegboard and hooks system. The LocHook is an innovative pegboard hook which can be mounted in any direction and is rugged enough to support just about anything. Together, the LocBoard and LocHook will provide the perfect stainless steel pegboard solution. Imagine a clean, well-kept kitchen with every pan, ladle, knife and utensil conveniently hanging and within grasp at all times. Imagine every medical supply in a hospital being easily accessible at a moment's notice. That is the power of the stainless steel pegboard system from Triton products.
Ordering information: Limited lifetime warranty.