2-(Difluoromethoxy)-5-fluorobromobenzene 97%

Supplier: Ambeed
Warning

A607279-100MG A607279-250MG
77266-778EA 49 USD
77266-778 77266-780
2-(Difluoromethoxy)-5-fluorobromobenzene 97%
2-(Difluoromethoxy)-5-fluorobromobenzene

Caution: for Research Use Only

Formula: C₇H₄BrF₃O
MDL Number: MFCD09935620
CAS Number: 936249-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR