Propargyl α-D-mannopyranoside 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Propyn-1-yl α-D-mannoside, Propargyl ALPHA-D-mannopyranoside, 2-Propynyl α-D-mannopyranoside
A567537-100MG
A567537-250MG
A567537-50MG
77266-412EA
248
USD
77266-412
77266-414
77266-416
Propargyl α-D-mannopyranoside 98%
Propargyl α-D-mannopyranoside
Caution: for Research Use Only
Formula:
C₉H₁₄O₆ |
MDL Number:
MFCD19980990 CAS Number: 854262-01-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...