4-Chloro-2-(trifluoromethyl)quinazoline 98%

Supplier: Ambeed
Warning

A470647-250MG
77265-336EA 19 USD
77265-336
4-Chloro-2-(trifluoromethyl)quinazoline 98%
4-Chloro-2-(trifluoromethyl)quinazoline

Caution: for Research Use Only

Formula: C₉H₄ClF₃N₂
MDL Number: MFCD00793683
CAS Number: 52353-35-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR