5(6)-Aminofluorescein (Mixture of isomers) 98% + mix
Supplier: Ambeed
|
Warning
|
Synonyms:
Fluoresceinamine, 5(or 6)-Amino-3′,6′-dihydroxyspiro[isobenzofuran-1(3H),9′-[9H]xanthen]-3-one, 5(6)-Aminofluorescein
A446964-1G
A446964-250MG
A446964-5G
77265-056EA
81
USD
77265-056
77265-058
77265-060
5(6)-Aminofluorescein (Mixture of isomers) 98% + mix
5(6)-Aminofluorescein (Mixture of isomers)
Caution: for Research Use Only
Formula:
C₂₀H₁₃NO₅ |
MDL Number:
MFCD00133331 CAS Number: 27599-63-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...