5(6)-Aminofluorescein (Mixture of isomers) 98% + mix

Supplier: Ambeed
Warning

Synonyms: Fluoresceinamine, 5(or 6)-Amino-3′,6′-dihydroxyspiro[isobenzofuran-1(3H),9′-[9H]xanthen]-3-one, 5(6)-Aminofluorescein

A446964-1G A446964-250MG A446964-5G
77265-056EA 81 USD
77265-056 77265-058 77265-060
5(6)-Aminofluorescein (Mixture of isomers) 98% + mix
5(6)-Aminofluorescein (Mixture of isomers)

Caution: for Research Use Only

Formula: C₂₀H₁₃NO₅
MDL Number: MFCD00133331
CAS Number: 27599-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR