4-Ethynyltoluene 98%

Supplier: Ambeed
Danger

Synonyms: 1-Ethynyl-4-methylbenzene, p-Tolylacetylene

A430229-10G A430229-1G A430229-25G A430229-5G
77264-848EA 21 USD
77264-848 77264-850 77264-852 77264-854
4-Ethynyltoluene 98%
4-Ethynyltoluene

Caution: for Research Use Only

Formula: C₉H₈
MW: 116.16 g/mol
MDL Number: MFCD00008571
CAS Number: 766-97-2
UN: 3295
ADR: 3 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR