Cysteamine 95%

Supplier: Ambeed
Danger

Synonyms: 2-Aminoethanethiol

A381010-1MG
77264-240EA 5.15 USD
77264-240
Cysteamine 95%
Cysteamine

Caution: for Research Use Only

Formula: C₂H₇NS
MW: 77.15 g/mol
MDL Number: MFCD00008196
CAS Number: 60-23-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR