2-Chlorobenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: (2-Chlorophenyl)methanol

A273523-100G A273523-10G A273523-25G A273523-500G
77262-528EA 27 USD
77262-528 77262-530 77262-532 77262-534
2-Chlorobenzyl alcohol 98%
2-Chlorobenzyl alcohol

Caution: for Research Use Only

Formula: C₇H₇ClO
MW: 142.58 g/mol
Boiling Pt: 227
MDL Number: MFCD00004604
CAS Number: 17849-38-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR