7-Isoquinolinol 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
7-Hydroxyisoquinoline, Isoquinolin-7-ol
A270646-1G
A270646-5G
77262-446EA
12.36
USD
77262-446
77262-448
77262-450
7-Isoquinolinol 95%
7-Isoquinolinol
Caution: for Research Use Only
Formula:
C₉H₇NO |
MDL Number:
MFCD00456131 CAS Number: 7651-83-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...