7-Isoquinolinol 95%

Supplier: Ambeed
Warning

Synonyms: 7-Hydroxyisoquinoline, Isoquinolin-7-ol

A270646-1G A270646-5G
77262-446EA 12.36 USD
77262-446 77262-448 77262-450
7-Isoquinolinol 95%
7-Isoquinolinol

Caution: for Research Use Only

Formula: C₉H₇NO
MDL Number: MFCD00456131
CAS Number: 7651-83-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR