Tetraethylene glycol monomethyl ether 98%

Supplier: Ambeed
Warning

Synonyms: mPEG4-OH, 2,5,8,11-Tetraoxatridecan-13-ol

A240186-25G A240186-100G A240186-10G A240186-500G A240186-1G A240186-5G A240186-1000G
103811-336EA 6 USD
103811-336 103851-134 77090-282 77261-764 77261-766 77261-768 77304-186 77304-188 77746-358
Tetraethylene glycol monomethyl ether 98%
Tetraethylene glycol monomethyl ether

Caution: for Research Use Only

Formula: C₉H₂₀O₅
MW: 208.25 g/mol
Boiling Pt: 285.4 °C (760 mm Hg)
Melting Pt: 39 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00041756
CAS Number: 23783-42-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR