2-Chloro-5-(trifluoromethyl)pyridine-4-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Chloro-5-(trifluoromethyl)isonicotinic acid
A211509-1G
A211509-5G
A211509-100G
A211509-10G
A211509-250MG
103818-112EA
8
USD
103818-112
103848-806
77087-216
77087-218
77261-062
77261-064
77261-066
77261-068
2-Chloro-5-(trifluoromethyl)pyridine-4-carboxylic acid 98%
2-Chloro-5-(trifluoromethyl)pyridine-4-carboxylic acid
Caution: for Research Use Only
Formula:
C₇H₃ClF₃NO₂ Boiling Pt: 281.6 °C (760 mm Hg) Melting Pt: 185…186 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01862019 CAS Number: 505084-58-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...