5-Methylbenzo[b]thiophene 98%

Supplier: Ambeed
Warning

A177623-100MG A177623-1G A177623-250MG A177623-5G
77260-134EA 6 USD
77260-134 77260-136 77260-138 77260-140
5-Methylbenzo[b]thiophene 98%
5-Methylbenzo[b]thiophene

Caution: for Research Use Only

Formula: C₉H₈S
MW: 148.23
MDL Number: MFCD00052509
CAS Number: 14315-14-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR