Allylboronic acid pinacol ester 97% stabilised

Supplier: Ambeed
Danger

Synonyms: 2-Allyl-4,4,5,5-Tetramethyl-1,3,2-Dioxaborolane

Stabilization: Stabilised with phenothiazine

A115496-10G A115496-25G A115496-1G A115496-5G
103798-576EA 9 USD
103798-576 103824-432 77257-926 77257-928 77297-374 77311-250
Allylboronic acid pinacol ester 97% stabilised
Allylboronic acid pinacol ester

Caution: for Research Use Only

Formula: C₉H₁₇BO₂
MW: 168.05 g/mol
Boiling Pt: 131.6 °C (760 mm Hg)
Flash Pt: 33.4 °C
Storage Temperature: Freezer
MDL Number: MFCD00013347
CAS Number: 72824-04-5
UN: 1993
ADR: 3 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR