4-Chloro-8-fluoroquinoline 97%

Supplier: Ambeed
Warning

A100892-250MG A100892-1G A100892-5G
103795-680EA 21 USD
103795-680 103821-370 77257-224 77257-226 77296-892 77296-894
4-Chloro-8-fluoroquinoline 97%
4-Chloro-8-fluoroquinoline

Caution: for Research Use Only

Formula: C₉H₅ClFN
Boiling Pt: 259.9 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00278785
CAS Number: 63010-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR