2,2,2-Tribromoethanol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Tribromoethyl alcohol
A749399-25G
A749399-5G
A749399-100G
A749399-10G
A749399-1MG
A749399-500G
103840-844EA
30
USD
103840-844
76845-074
76845-076
77255-580
77255-582
77255-584
77747-477
2,2,2-Tribromoethanol 97%
2,2,2-Tribromoethanol
Caution: for Research Use Only
Formula:
Br₃CCH₂OH Boiling Pt: 199 °C (760 mm Hg) Melting Pt: 73…79 °C (760 mm Hg) Flash Pt: 99.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004671 CAS Number: 75-80-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...