2,3,7,8,12,13,17,18-Octaethylporphyrin 97%

Supplier: Ambeed
Warning

Synonyms: OEP, Octaethylporphine, 2,3,7,8,12,13,17,18-Octaethyl-21H,23H-porphine

A613242-1G
77254-340EA 284.28 USD
77254-340
2,3,7,8,12,13,17,18-Octaethylporphyrin 97%
2,3,7,8,12,13,17,18-Octaethylporphyrin

Caution: for Research Use Only

Formula: C₃₆H₄₆N₄
MW: 534.79 g/mol
MDL Number: MFCD00009627
CAS Number: 2683-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR