cis-Dichlorobis(triethylphosphine)platinum(II) 98%

Supplier: Ambeed
Warning

A552715-100MG A552715-1G A552715-250MG
77253-848EA 46 USD
77253-848 77253-850 77253-852
cis-Dichlorobis(triethylphosphine)platinum(II) 98%
cis-Dichlorobis(triethylphosphine)platinum(II)

Caution: for Research Use Only

Formula: PtCl₂[P(C₂H₅)₃]₂
MW: 502.304 g/mol
MDL Number: MFCD00058821
CAS Number: 15692-07-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR