cis-Tetrakis(dimethylsulfoxide)dichlororuthenium(II) 98%

Supplier: Ambeed
Danger

Synonyms: cis-Dichlorotetrakis(dimethyl sulphoxide)ruthenium, cis-Dichlorotetrakis(dimethyl sulfoxide)ruthenium

A434326-100MG A434326-1G A434326-250MG A434326-5G
77252-488EA 44 USD
77252-488 77252-490 77252-492 77252-494
cis-Tetrakis(dimethylsulfoxide)dichlororuthenium(II) 98%
cis-Tetrakis(dimethylsulfoxide)dichlororuthenium(II)

Caution: for Research Use Only

Formula: C₈H₂₄Cl₂O₄RuS₄
MDL Number: MFCD00050955
CAS Number: 59091-96-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR