Tris[triphenylphosphinegold(I)]oxonium tetrafluoroborate 99% Au 39.91 ± 0.8%
Supplier: Ambeed
|
Warning
|
A429915-100MG
A429915-1G
A429915-250MG
A429915-5G
77252-436EA
100
USD
77252-436
77252-438
77252-440
77252-442
Tris[triphenylphosphinegold(I)]oxonium tetrafluoroborate 99% Au 39.91 ± 0.8%
Tris[triphenylphosphinegold(I)]oxonium tetrafluoroborate
Caution: for Research Use Only
Formula:
C₅₄H₄₈Au₃B₄F₁₆OP₃ |
MDL Number:
MFCD06658141 CAS Number: 53317-87-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...