2,7-Dichlorofluorene 97%

Supplier: Ambeed
Warning

Synonyms: 2,7-Dichloro-9H-fluorene

A411926-100G A411926-1G A411926-25G A411926-500G A411926-5G
77252-238EA 55 USD
77252-238 77252-240 77252-242 77252-244 77252-246
2,7-Dichlorofluorene 97%
2,7-Dichlorofluorene

Caution: for Research Use Only

Formula: C₁₃H₈Cl₂
MDL Number: MFCD00032840
CAS Number: 7012-16-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR