Fluorescent brightener 378 97%

Supplier: Ambeed
Warning

A369900-25G A369900-100G
77251-748EA 12 USD
77251-748 77746-745
Fluorescent brightener 378 97%
Fluorescent brightener 378

Caution: for Research Use Only

Formula: C₆₀H₅₂O₄
MDL Number: MFCD03425832
CAS Number: 164908-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR