2,2-Dimethylcyclopentanone 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
2,2-Dimethylcyclopentan-1-one
A324989-100MG
A324989-1G
A324989-250MG
A324989-5G
77251-194EA
5
USD
77251-194
77251-196
77251-198
77251-200
2,2-Dimethylcyclopentanone 97%
2,2-Dimethylcyclopentanone
Caution: for Research Use Only
Formula:
C₇H₁₂O |
MDL Number:
MFCD00049164 CAS Number: 4541-32-6 UN: 1224 ADR: 3,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...