2,2-Dimethylcyclopentanone 97%

Supplier: Ambeed
Danger

Synonyms: 2,2-Dimethylcyclopentan-1-one

A324989-100MG A324989-1G A324989-250MG A324989-5G
77251-194EA 5 USD
77251-194 77251-196 77251-198 77251-200
2,2-Dimethylcyclopentanone 97%
2,2-Dimethylcyclopentanone

Caution: for Research Use Only

Formula: C₇H₁₂O
MDL Number: MFCD00049164
CAS Number: 4541-32-6
UN: 1224
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR