(2S)-(-)-3,3'-Diphenyl-[2,2'-binaphthalene]-1,1'-diol ≥98%, ee 99%

Supplier: Ambeed
Danger

A238090-100MG A238090-250MG A238090-50MG A238090-5G
77249-704EA 70 USD
77249-704 77249-706 77249-708 77249-710
(2S)-(-)-3,3'-Diphenyl-[2,2'-binaphthalene]-1,1'-diol ≥98%, ee 99%
(2S)-(-)-3,3'-Diphenyl-[2,2'-binaphthalene]-1,1'-diol

Caution: for Research Use Only

Formula: C₃₂H₂₂O₂
MDL Number: MFCD08459338
CAS Number: 147702-14-5
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR