2-(Diphenylphosphino)benzaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: (2-Formylphenyl)diphenylphosphine

A195130-1G A195130-100G A195130-10G A195130-250MG A195130-500G A195130-25G A195130-5G
103816-726EA 6 USD
103816-726 103847-340 77085-294 77248-660 77248-662 77248-664 77248-666 77302-446 77302-448
2-(Diphenylphosphino)benzaldehyde 97%
2-(Diphenylphosphino)benzaldehyde

Caution: for Research Use Only

Formula: C₁₉H₁₅OP
Boiling Pt: 417.4 °C (760 mm Hg)
Melting Pt: 112…115 °C
Flash Pt: 206.263 °C
Storage Temperature: Ambient
MDL Number: MFCD00013367
CAS Number: 50777-76-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR