2,4,6-Tri-tert-butylaniline 98%

Supplier: Ambeed
Warning

Synonyms: 2,4,6-Tris(tert-butyl)aniline

A178570-5G A178570-100G A178570-100MG A178570-10G A178570-250MG A178570-25G
103807-354EA 14 USD
103807-354 103845-690 77083-122 77248-200 77248-202 77248-204 77248-206 77301-564
2,4,6-Tri-tert-butylaniline 98%
2,4,6-Tri-tert-butylaniline

Caution: for Research Use Only

Formula: C₁₈H₃₁N
Boiling Pt: 302 °C (760 mm Hg)
Melting Pt: 145…147 °C
Flash Pt: 123.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00011645
CAS Number: 961-38-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR