2-Adamantylamine hydrochloride 97%

Supplier: Ambeed
Warning

A166662-1G A166662-250MG A166662-25G
77247-870EA 16 USD
77247-870 77247-872 77567-424
2-Adamantylamine hydrochloride 97%
2-Adamantylamine hydrochloride

Caution: For research use only.

Formula: C₁₀H₁₈ClN
MDL Number: MFCD00074743
CAS Number: 10523-68-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR