(S)-(+)-1-[(R)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldiphenylphosphine 98%

Supplier: Ambeed
Warning

Synonyms: (S)-1-[(S)-2-(Dicyclohexylphosphino)ferrocenylethyl]diphenylphosphine

A161611-100MG A161611-250MG A161611-1G
77247-674EA 13 USD
77247-674 77247-676 77550-026
(S)-(+)-1-[(R)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldiphenylphosphine 98%
(S)-(+)-1-[(R)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldiphenylphosphine

Caution: for Research Use Only

Formula: C₃₆H₄₄FeP₂
MDL Number: MFCD00800281
CAS Number: 162291-01-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR