N-Benzylglycine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Bzl-Gly-OH·HCl, Bzl-Gly-OH.HCl

A151351-100G A151351-10G A151351-25G A151351-5G
77247-266EA 73 USD
77247-266 77247-268 77247-270 77247-272
N-Benzylglycine hydrochloride 98%
N-Benzylglycine hydrochloride

Caution: for Research Use Only

Formula: C₉H₁₁NO₂·HCl
MW: 201.7 g/mol
MDL Number: MFCD00156934
CAS Number: 7689-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR