Luteolin 98%
Supplier: Ambeed
|
Warning
|
A144892-100G
A144892-10G
A144892-500G
A144892-1G
A144892-25G
A144892-5G
103803-466EA
6
USD
103803-466
103841-544
77247-090
77247-092
77247-094
77299-420
77299-422
77299-424
Luteolin 98%
Luteolin
Caution: for Research Use Only
Formula:
C₁₅H₁₀O₆ Melting Pt: 330 °C Flash Pt: 239.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00017309 CAS Number: 491-70-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...